N-Methyl-4-nitrophthalimide ≥99.0% (by HPLC)

Supplier: TCI America

Synonyms: 2-Methyl-5-nitroisoindoline-1,3-dione

M1310-25G M1310-5G
TCM1310-025GEA 240.16 CAD
TCM1310-025G TCM1310-005G
N-Methyl-4-nitrophthalimide ≥99.0% (by HPLC)
N-Methyl-4-nitrophthalimide

CAS Number: 41663-84-7
MDL Number: MFCD00454263
Molecular Formula: C9H6N2O4
Molecular Weight: 206.16
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 178

Formula: C₉H₆N₂O₄
MW: 206.16 g/mol
Melting Pt: 178 °C
MDL Number: MFCD00454263
CAS Number: 41663-84-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=518f497d0e2a3d9b27f5d523b926aea5f25c9ebb&vid=518f497d0e2a3d9b27f5d523b926aea5f25c9ebb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR