3-Methoxymethylindole ≥98.0%

Supplier: TCI America

M1305-5G M1305-25G M1305-1G
TCM1305-5GEA 532.46 CAD
TCM1305-5G TCM1305-025G TCM1305-1G
3-Methoxymethylindole ≥98.0%
3-Methoxymethylindole

CAS Number: 78440-76-3
MDL Number: MFCD00276420
Molecular Formula: C10H11NO
Molecular Weight: 161.20
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal

Formula: C₁₀H₁₁NO
MW: 161.20 g/mol
MDL Number: MFCD00276420
CAS Number: 78440-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf21d9c27e7c505071d26d7ff93991af9b53c57b&vid=bf21d9c27e7c505071d26d7ff93991af9b53c57b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR