2-Methyl-4,5-diphenyloxazole

Supplier: TCI America

M1299-5G M1299-25G
TCM1299-005GEA 157.05 CAD
TCM1299-005G TCM1299-025G
2-Methyl-4,5-diphenyloxazole
2-Methyl-4,5-diphenyloxazole

CAS Number: 14224-99-8
MDL Number: MFCD00005307
Molecular Formula: C16H13NO
Molecular Weight: 235.29
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 212
Flash Point (°C): 113
Specific Gravity (20/20): 1.14

Formula: C₁₆H₁₃NO
Boiling Pt: 212 °C
Density: 1.14 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00005307
CAS Number: 14224-99-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7602cc2ece4937122988a9fd149ab0bd65514ba&vid=a7602cc2ece4937122988a9fd149ab0bd65514ba&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR