Ethyl-6-methylsalicylate ≥98.0%

Supplier: TCI America

M1283-25G M1283-5G
TCM1283-25GEA 1148.82 CAD
TCM1283-25G TCM1283-005G
Ethyl-6-methylsalicylate ≥98.0%
Ethyl-6-methylsalicylate

CAS Number: 6555-40-4
MDL Number: MFCD01194299
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 45


Other Product Names / Synonyms: Ethyl 2-Hydroxy-6-methylbenzoate, 2-Hydroxy-6-methylbenzoic Acid Ethyl Ester, 6-Methylsalicylic Acid Ethyl Ester

Formula: C₁₀H₁₂O₃
Melting Pt: 45 °C
MDL Number: MFCD01194299
CAS Number: 6555-40-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc44c0465ba2ac5b7e512a34e194e6c29695e582&vid=dc44c0465ba2ac5b7e512a34e194e6c29695e582&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR