1-(Methoxymethyl)benzotriazole ≥98.0%
Supplier: TCI America
M1276-1G
M1276-5G
TCM1276-001GEA
127.35
CAD
TCM1276-001G
TCM1276-005G
1-(Methoxymethyl)benzotriazole ≥98.0%
1-(Methoxymethyl)benzotriazole
CAS Number: 71878-80-3
MDL Number: MFCD00216630
Molecular Formula: C8H9N3O
Molecular Weight: 163.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 40
Formula:
C₈H₉N₃O Melting Pt: 40 °C |
MDL Number:
MFCD00216630 CAS Number: 71878-80-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...