(S)-(+)-Ethyl mandelate ≥98.0%
Supplier: TCI America
Synonyms:
L-(+)-Mandelic acid ethyl ester, (S)-Ethyl 2-hydroxy-2-phenylacetate, Ethyl L-(+)-Mandelate
CAS Number: 13704-09-1
MDL Number: MFCD00064249
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 107
Flash Point (°C): 110
Freezing point (°C): 28
Specific rotation [a]20/D: 137 deg (C=1, CHCl3)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: L-(+)-Mandelic Acid Ethyl Ester
Formula:
C₁₀H₁₂O₃ MW: 180.2 g/mol Boiling Pt: 107 °C Flash Pt: 110 °C |
MDL Number:
MFCD00064249 CAS Number: 13704-09-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...