1-(2-Mesitylene)-1,3-butanedione ≥96.0%
Supplier: TCI America
Synonyms:
(2,4,6-Trimethylbenzoyl)acetone, 1-(2,4,6-Trimethylbenzene)-1,3-butanedione
M1272-5G
M1272-25G
TCM1272-005GEA
360.24
CAD
TCM1272-005G
TCM1272-25G
1-(2-Mesitylene)-1,3-butanedione ≥96.0%
1-(2-Mesitylene)-1,3-butanedione
CAS Number: 6450-57-3
MDL Number: MFCD00477603
Molecular Formula: C13H16O2
Molecular Weight: 204.27
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 172
Melting point (°C): 45
Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 1-(2,4,6-Trimethylbenzene)-1,3-butanedione, (2,4,6-Trimethylbenzoyl)acetone
Formula:
C₁₃H₁₆O₂ Boiling Pt: 172 °C Melting Pt: 45 °C |
MDL Number:
MFCD00477603 CAS Number: 6450-57-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...