2-Methoxybutyl acetate ≥97.0%
Supplier: TCI America
Synonyms:
Acetic Acid 2-Methoxybutyl Ester
A1675-25G
A1675-5G
TCA1675-25GEA
794.74
CAD
TCA1675-25G
TCA1675-5G
2-Methoxybutyl acetate ≥97.0%
2-Methoxybutyl acetate
CAS Number: 1173168-18-7
MDL Number: MFCD06797047
Molecular Formula: C7H14O3
Molecular Weight: 146.19
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Other Product Names / Synonyms: Acetic Acid 2-Methoxybutyl Ester
Formula:
C₇H₁₄O₃ MW: 146.19 g/mol |
MDL Number:
MFCD06797047 CAS Number: 1173168-18-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...