2-(Methylthio)-2-thiazoline ≥98.0%

Supplier: TCI America

Synonyms: 2-(Methylmercapto)-2-thiazoline, 2-(Methylthio)-4,5-dihydrothiazole

M1262-25G M1262-5G
TCM1262-025GEA 398.32 CAD
TCM1262-025G TCM1262-5G
2-(Methylthio)-2-thiazoline ≥98.0%
2-(Methylthio)-2-thiazoline

CAS Number: 19975-56-5
MDL Number: MFCD00005312
Molecular Formula: C4H7NS2
Molecular Weight: 133.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 217
Flash Point (°C): 97
Specific Gravity (20/20): 1.23


Other Product Names / Synonyms: 2-(Methylmercapto)-2-thiazoline

Formula: C₄H₇NS₂
MW: 133.23 g/mol
Boiling Pt: 217 °C
Density: 1.23 (20 °C)
Flash Pt: 97 °C
MDL Number: MFCD00005312
CAS Number: 19975-56-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94ec1a1cb84abc40a9e27eecc1bc02658320b94c&vid=94ec1a1cb84abc40a9e27eecc1bc02658320b94c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR