2-Methoxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
M1261-25G
M1261-1G
M1261-5G
TCM1261-25GEA
127.98
CAD
TCM1261-25G
TCM1261-001G
TCM1261-005G
2-Methoxyphenylboronic acid (contains varying amounts of Anhydride)
2-Methoxyphenylboronic acid
CAS Number: 5720-06-9
MDL Number: MFCD00236047
Molecular Formula: C7H9BO3
Molecular Weight: 151.96
Form: Crystal
Color: White
Melting point (°C): 108
Other Product Names / Synonyms: 2-Methoxybenzeneboronic Acid
Formula:
C₇H₉BO₃ Melting Pt: 108 °C |
MDL Number:
MFCD00236047 CAS Number: 5720-06-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...