2,2'-Methylenebis[6-(benzotriazol-2-yl)-4-tert-octylphenol] ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Bis[3-(benzotriazol-2-yl)-2-hydroxy-5-tert-octylphenyl]methane

M1259-25G M1259-500G
TCM1259-025GEA 85.95 CAD
TCM1259-025G TCM1259-500G
2,2'-Methylenebis[6-(benzotriazol-2-yl)-4-tert-octylphenol] ≥98.0% (by HPLC)
2,2'-Methylenebis[6-(benzotriazol-2-yl)-4-tert-octylphenol]

CAS Number: 103597-45-1
MDL Number: MFCD00192289
Molecular Formula: C41H50N6O2
Molecular Weight: 658.89
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 198
Flash Point (°C): 200


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Bis[3-(benzotriazol-2-yl)-2-hydroxy-5-tert-octylphenyl]methane

Formula: C₄₁H₅₀N₆O₂
Melting Pt: 198 °C
Flash Pt: 200 °C
MDL Number: MFCD00192289
CAS Number: 103597-45-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65be0315a292f68ae36f035b13316a9b62a0b0a5&vid=85d5e4c80eb3daf61b95bd53e8d3f6ca9cec2769&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR