Methyl-β-D-xylopyranoside ≥96.0%
Supplier: TCI America
M1253-25G
M1253-5G
TCM1253-025GEA
388.68
CAD
TCM1253-025G
TCM1253-005G
Methyl-β-D-xylopyranoside ≥96.0%
Methyl-beta-D-xylopyranoside
CAS Number: 612-05-5
MDL Number: MFCD00047532
Molecular Formula: C6H12O5
Molecular Weight: 164.16
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 155
Specific rotation [a]20/D: -63 deg (C=2, H2O)
Formula:
C₆H₁₂O₅ Melting Pt: 155 °C |
MDL Number:
MFCD00047532 CAS Number: 612-05-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...