2-Ethoxyethyl methacrylate ≥98.0% stabilised for synthesis

Supplier: TCI America

Synonyms: Methacrylic acid 2-Ethoxyethyl ester, Ethylene glycol monoethyl ether Methacrylate

M1251-25ML M1251-500ML
TCM1251-025MLEA 47.4 CAD
TCM1251-025ML TCM1251-500ML
2-Ethoxyethyl methacrylate ≥98.0% stabilised for synthesis
2-Ethoxyethyl methacrylate

(stabilized with MEHQ)
CAS Number: 2370-63-0
MDL Number: MFCD00009249
Molecular Formula: C8H14O3
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 93
Flash Point (°C): 72
Specific Gravity (20/20): 0.97


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Ethylene Glycol Monoethyl Ether Methacrylate, Methacrylic Acid 2-Ethoxyethyl Ester

Formula: C₈H₁₄O₃
MW: 158.1950 g/mol
Boiling Pt: 93 °C
Density: 0.97 (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00009249
CAS Number: 2370-63-0
EINECS: 219-135-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=543352a1803af3bf7e8b4534d1bee3fb8a269249&vid=543352a1803af3bf7e8b4534d1bee3fb8a269249&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR