2-(4-Methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine ≥98.0%
Supplier: TCI America
Synonyms:
4,6-Bis(trichloromethyl)-2-(4-methoxyphenyl)-1,3,5-triazine
M1245-5G
M1245-25G
TCM1245-005GEA
118.93
CAD
TCM1245-005G
TCM1245-25G
2-(4-Methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine ≥98.0%
2-(4-Methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine
CAS Number: 3584-23-4
MDL Number: MFCD00521486
Molecular Formula: C12H7Cl6N3O
Molecular Weight: 421.91
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 145
Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: 4,6-Bis(trichloromethyl)-2-(4-methoxyphenyl)-1,3,5-triazine
Formula:
C₁₂H₇Cl₆N₃O MW: 421.91 g/mol Melting Pt: 145 °C |
MDL Number:
MFCD00521486 CAS Number: 3584-23-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...