4-Amino-4'-dimethylaminoazobenzene ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

A1672-5G A1672-1G
TCA1672-5GEA 661.23 CAD
TCA1672-5G TCA1672-1G
4-Amino-4'-dimethylaminoazobenzene ≥97.0% (by HPLC, titration analysis)
4-Amino-4'-dimethylaminoazobenzene

CAS Number: 539-17-3
MDL Number: MFCD00010204
Molecular Formula: C14H16N4
Molecular Weight: 240.31
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 189


Other Product Names / Synonyms: N,N-Dimethyl-4,4'-azodianiline

Formula: C₁₄H₁₆N₄
Melting Pt: 189 °C
MDL Number: MFCD00010204
CAS Number: 539-17-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa09040365b0389a5a489a0ade94097a402e2f8a&vid=aa09040365b0389a5a489a0ade94097a402e2f8a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR