Ethyl-4-methoxycinnamate ≥98.0%

Supplier: TCI America

Synonyms: 4-Methoxycinnamic Acid Ethyl Ester, (E)-Ethyl p-methoxycinnamate, (E)-Ethyl 3-(4-methoxyphenyl)acrylate, Ethyl 3-(4-Methoxyphenyl)acrylate, 3-(4-Methoxyphenyl)acrylic Acid Ethyl Ester

M1204-5G M1204-25G
TCM1204-5GEA 118.97 CAD
TCM1204-5G TCM1204-025G
Ethyl-4-methoxycinnamate ≥98.0%
Ethyl-4-methoxycinnamate

CAS Number: 24393-56-4
MDL Number: MFCD00026906
Molecular Formula: C12H14O3
Molecular Weight: 206.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 187
Melting point (°C): 49


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 4-Methoxycinnamic Acid Ethyl Ester, Ethyl 3-(4-Methoxyphenyl)acrylate, 3-(4-Methoxyphenyl)acrylic Acid Ethyl Ester

Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
Boiling Pt: 187 °C
Melting Pt: 49 °C
MDL Number: MFCD00026906
CAS Number: 24393-56-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f681a453f9cfd4c7c809e1246b50247994da125&vid=6f681a453f9cfd4c7c809e1246b50247994da125&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR