4-Methyl-1-acetonaphthone ≥97.0%

Supplier: TCI America

Synonyms: 1-Acetyl-4-methylnaphthalene, 1-(4-Methylnaphthalen-1-yl)ethanone

M1202-5G M1202-25G
TCM1202-005GEA 45.82 CAD
TCM1202-005G TCM1202-25G
4-Methyl-1-acetonaphthone ≥97.0%
4-Methyl-1-acetonaphthone

CAS Number: 28418-86-2
MDL Number: MFCD00671556
Molecular Formula: C13H12O
Molecular Weight: 184.24
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 136
Melting point (°C): 39


Other Product Names / Synonyms: 1-Acetyl-4-methylnaphthalene

Formula: C₁₃H₁₂O
MW: 184.24 g/mol
Boiling Pt: 136 °C
Melting Pt: 39 °C
MDL Number: MFCD00671556
CAS Number: 28418-86-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8381963943cf669c8f53e2655a911352d7426754&vid=8381963943cf669c8f53e2655a911352d7426754&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR