3-Methyl-4-cyclohexene-1,2-dicarboxylic anhydride ≥95.0% (by titrimetric analysis)

Supplier: TCI America

M1190-1G M1190-5G
TCM1190-001GEA 325.48 CAD
TCM1190-001G TCM1190-5G
3-Methyl-4-cyclohexene-1,2-dicarboxylic anhydride ≥95.0% (by titrimetric analysis)
3-Methyl-4-cyclohexene-1,2-dicarboxylic anhydride

CAS Number: 5333-84-6
MDL Number: MFCD00014585
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 62


Other Product Names / Synonyms: 3-Methyl-DELTA4-tetrahydrophthalic Anhydride

Formula: C₉H₁₀O₃
Melting Pt: 62 °C
MDL Number: MFCD00014585
CAS Number: 5333-84-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b46ee43ad350598c373c916caeed1573ad0745fe&vid=b46ee43ad350598c373c916caeed1573ad0745fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR