Sodium-3-mercapto-1-propanesulfonate ≥85.0%

Supplier: TCI America

Synonyms: Sodium 3-mercaptopropanesulphonate, 3-Mercapto-1-propanesulphonic acid Sodium Salt, Sodium 3-mercaptopropanesulfonate, 3-Mercapto-1-propanesulfonic acid Sodium Salt

M1189-25G M1189-500G M1189-100G
TCM1189-025GEA 52.93 CAD
TCM1189-025G TCM1189-500G TCM1189-100G
Sodium-3-mercapto-1-propanesulfonate ≥85.0%
Sodium-3-mercapto-1-propanesulfonate

CAS Number: 17636-10-1
MDL Number: MFCD00043148
Molecular Formula: C3H8O3S2
Molecular Weight: 178.20
Purity/Analysis Method: >85.0% (T)
Form: Crystal
Melting point (°C): 220


Other Product Names / Synonyms: 3-Mercapto-1-propanesulfonic Acid Sodium Salt

Formula: C₃H₈O₃S₂
Melting Pt: 220 °C
MDL Number: MFCD00043148
CAS Number: 17636-10-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90be71ceae0b387f6556df9f028b281f92bc26a6&vid=90be71ceae0b387f6556df9f028b281f92bc26a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR