N-Methyl-N-(2-pyridyl)formamide ≥98.0%

Supplier: TCI America

M1187-1G M1187-5G
TCM1187-001GEA 75.84 CAD
TCM1187-001G TCM1187-005G
N-Methyl-N-(2-pyridyl)formamide ≥98.0%
N-Methyl-N-(2-pyridyl)formamide

CAS Number: 67242-59-5
MDL Number: MFCD00006260
Molecular Formula: C7H8N2O
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Flash Point (°C): 109
Specific Gravity (20/20): 1.16


Product Categories: Synthetic Organic Chemistry, Formylation
Other Product Names / Synonyms: 2-(N-Formyl-N-methylamino)pyridine, Meyer's Reagent

Formula: C₇H₈N₂O
Boiling Pt: 82 °C
Density: 1.16 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00006260
CAS Number: 67242-59-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b73ed2090eb7f33ecbb71ffafaa7ef686aa2e03c&vid=b73ed2090eb7f33ecbb71ffafaa7ef686aa2e03c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR