(S)-(-)-1-Methyl-2-(1-piperidinomethyl)pyrrolidine ≥97.0% (by GC, titration analysis)
Supplier: TCI America
M1183-1G
M1183-5G
TCM1183-001GEA
55.3
CAD
TCM1183-001G
TCM1183-005G
(S)-(-)-1-Methyl-2-(1-piperidinomethyl)pyrrolidine ≥97.0% (by GC, titration analysis)
(S)-(-)-1-Methyl-2-(1-piperidinomethyl)pyrrolidine
CAS Number: 84466-85-3
MDL Number: MFCD00077821
Molecular Formula: C11H22N2
Molecular Weight: 182.31
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 126
Flash Point (°C): 105
Specific Gravity (20/20): 0.91
Specific rotation [a]20/D: -69 deg (C=0.5, EtOH)
Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts
Other Product Names / Synonyms: (S)-(-)-1-[(1-Methyl-2-pyrrolidinyl)methyl]piperidine
Formula:
C₁₁H₂₂N₂ Boiling Pt: 126 °C Density: 0.91 (20 °C) Flash Pt: 105 °C |
MDL Number:
MFCD00077821 CAS Number: 84466-85-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...