Ethyl-O-Mesitylsulfonylacetohydroxamate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl N-(mesitylsulfonyl)oxyacetimidate, O-Mesitylsulphonylacetohydroxamic acid Ethyl Ester, Ethyl N-(mesitylsulphonyl)oxyacetimidate, O-Mesitylsulfonylacetohydroxamic acid Ethyl Ester

M1182-1G M1182-5G M1182-25G
TCM1182-1GEA 52.14 CAD
TCM1182-1G TCM1182-005G TCM1182-25G
Ethyl-O-Mesitylsulfonylacetohydroxamate ≥98.0%
Ethyl-O-Mesitylsulfonylacetohydroxamate

[Precursor of the Powerful Aminating Reagent]
CAS Number: 38202-27-6
MDL Number: MFCD00009244
Molecular Formula: C13H19NO4S
Molecular Weight: 285.36
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 56
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Amination
Other Product Names / Synonyms: O-Mesitylsulfonylacetohydroxamic Acid Ethyl Ester

Formula: C₁₃H₁₉NO₄S
Melting Pt: 56 °C
MDL Number: MFCD00009244
CAS Number: 38202-27-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=259b74dca527da4b245368a8ccf752bb8b943053&vid=259b74dca527da4b245368a8ccf752bb8b943053&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR