3-((4-Nitrobenzyl)oxy)-3-oxopropanoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
M1177-5G
M1177-25G
TCM1177-005GEA
160.37
CAD
TCM1177-005G
TCM1177-25G
3-((4-Nitrobenzyl)oxy)-3-oxopropanoic acid ≥98.0% (by titrimetric analysis)
3-((4-Nitrobenzyl)oxy)-3-oxopropanoic acid
CAS Number: 77359-11-6
MDL Number: MFCD00191556
Molecular Formula: C10H9NO6
Molecular Weight: 239.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 109
Other Product Names / Synonyms: Malonic Acid Mono-4-nitrobenzyl Ester, 4-Nitrobenzyl Hydrogen Malonate
Formula:
C₁₀H₉NO₆ Melting Pt: 109 °C |
MDL Number:
MFCD00191556 CAS Number: 77359-11-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...