N-Methyl-1-naphthylamine hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
M1172-5G
M1172-25G
TCM1172-5GEA
158
CAD
TCM1172-5G
TCM1172-025G
N-Methyl-1-naphthylamine hydrochloride ≥98.0% (by titrimetric analysis)
N-Methyl-1-naphthylamine hydrochloride
CAS Number: 4643-36-1
MDL Number: MFCD00191555
Molecular Formula: C11H11N
Molecular Weight: 193.67
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 177
Other Product Names / Synonyms: 1-(N-Methylamino)naphthalene Hydrochloride
Formula:
C₁₁H₁₁N Melting Pt: 177 °C |
MDL Number:
MFCD00191555 CAS Number: 4643-36-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...