4-Methoxy-3-nitrobiphenyl ≥99.0%

Supplier: TCI America

Synonyms: 2-Nitro-4-phenylanisole

M1166-25G M1166-5G
TCM1166-025GEA 150.1 CAD
TCM1166-025G TCM1166-5G
4-Methoxy-3-nitrobiphenyl ≥99.0%
4-Methoxy-3-nitrobiphenyl

CAS Number: 15854-73-6
MDL Number: MFCD00191554
Molecular Formula: C13H11NO3
Molecular Weight: 229.24
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 89


Other Product Names / Synonyms: 2-Nitro-4-phenylanisole

Formula: C₁₃H₁₁NO₃
MW: 229.24 g/mol
Melting Pt: 89 °C
MDL Number: MFCD00191554
CAS Number: 15854-73-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2c92c4188b7aa083f85d12a149567cdd3cc7f10&vid=d2c92c4188b7aa083f85d12a149567cdd3cc7f10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR