3-Sulfopropyl methacrylate potassium salt ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Potassium 3-(Methacryloyloxy)propanesulfonate, Methacrylic acid 3-sulphopropyl Ester Potassium salt, 3-sulphopropyl Methacrylate Potassium Salt, Methacrylic acid 3-sulfopropyl Ester Potassium salt, 3-sulfopropyl Methacrylate Potassium Salt, Potassium 3-(Methacryloyloxy)propanesulphonate

M1158-25G M1158-500G
TCM1158-025GEA 193.71 CAD
TCM1158-025G TCM1158-500G
3-Sulfopropyl methacrylate potassium salt ≥97.0% (by HPLC, titration analysis)
3-Sulfopropyl methacrylate potassium salt

CAS Number: 31098-21-2
MDL Number: MFCD00010139
Molecular Formula: C7H12O5S
Molecular Weight: 246.32
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 295


Product Categories: Materials Chemistry, Monomers, Bioscience, Monomers for Synthetic Polymers (for DDS)
Other Product Names / Synonyms: Potassium 3-(Methacryloyloxy)propanesulfonate, Methacrylic Acid 3-Sulfopropyl Ester Potassium Salt

Formula: C₇H₁₂O₅S
MW: 246.32 g/mol
Melting Pt: 295 °C
MDL Number: MFCD00010139
CAS Number: 31098-21-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3f805c55bb2eb3b7abf1b5b87d77ae8069d059a&vid=c3f805c55bb2eb3b7abf1b5b87d77ae8069d059a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR