Ethyl-4-methylpyrrole-2-carboxylate ≥97.0%

Supplier: TCI America

M1145-1G M1145-5G
TCM1145-001GEA 393.42 CAD
TCM1145-001G TCM1145-5G
Ethyl-4-methylpyrrole-2-carboxylate ≥97.0%
Ethyl 4-methylpyrrole-2-carboxylate

CAS Number: 40611-85-6
MDL Number: MFCD00191668
Molecular Formula: C8H11NO2
Molecular Weight: 153.18
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 104
Melting point (°C): 43


Other Product Names / Synonyms: 4-Methyl-2-pyrrolecarboxylic Acid Ethyl Ester

Formula: C₈H₁₁NO₂
Boiling Pt: 104 °C
Melting Pt: 43 °C
MDL Number: MFCD00191668
CAS Number: 40611-85-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d3bb6ec7dde585e6cf1b7751daf7722695852f3&vid=4d3bb6ec7dde585e6cf1b7751daf7722695852f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR