3-Methylpicolinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Methylpyridine-2-carboxylic acid

M1141-100MG M1141-1G M1141-5G
TCM1141-100MGEA 26.86 CAD
TCM1141-100MG TCM1141-001G TCM1141-5G
3-Methylpicolinic acid ≥98.0% (by GC, titration analysis)
3-Methylpicolinic acid

CAS Number: 4021-07-2
MDL Number: MFCD00191553
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 119


Other Product Names / Synonyms: 3-Methylpicolinic Acid

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 119 °C
MDL Number: MFCD00191553
CAS Number: 4021-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d04e27a3388b4e6a2866e98142e64975a390fee5&vid=d04e27a3388b4e6a2866e98142e64975a390fee5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR