4-(4-Aminopiperidino)pyridine dihydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Amino-1-(4-pyridyl)piperidine Dihydrochloride

A1655-1G A1655-5G
TCA1655-1GEA 118.97 CAD
TCA1655-1G TCA1655-5G
4-(4-Aminopiperidino)pyridine dihydrochloride ≥98.0% (by HPLC, titration analysis)
4-(4-Aminopiperidino)pyridine dihydrochloride

CAS Number: 1169396-92-2
MDL Number: MFCD06797043
Molecular Formula: C10H15N3
Molecular Weight: 250.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 4-Amino-1-(4-pyridyl)piperidine Dihydrochloride

Formula: C₁₀H₁₅N₃
MW: 250.17 g/mol
MDL Number: MFCD06797043
CAS Number: 1169396-92-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8eee6b747b0622796ebaef9bb93bf3fe448667c&vid=b8eee6b747b0622796ebaef9bb93bf3fe448667c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR