Dibutyl methylenebis(thioglycolate) ≥95.0%

Supplier: TCI America

M1137-25G M1137-500G
TCM1137-025GEA 60.04 CAD
TCM1137-025G TCM1137-500G
Dibutyl methylenebis(thioglycolate) ≥95.0%
Dibutyl methylenebis(thioglycolate)

CAS Number: 14338-82-0
MDL Number: MFCD00027216
Molecular Formula: C13H24O4S2
Molecular Weight: 308.45
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 178
Flash Point (°C): 167
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: Methylenebis(thioglycolic Acid Dibutyl Ester)

Formula: C₁₃H₂₄O₄S₂
Boiling Pt: 178 °C
Density: 1.09 (20 °C)
Flash Pt: 167 °C
MDL Number: MFCD00027216
CAS Number: 14338-82-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93540ae1aa8eaf9fb1b6bddc66e80d9b1be8b2c2&vid=93540ae1aa8eaf9fb1b6bddc66e80d9b1be8b2c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR