3-Methoxydiphenylamine ≥98.0%

Supplier: TCI America

Synonyms: 3-Methoxy-N-phenylaniline

M1135-25G M1135-500G
TCM1135-025GEA 55.3 CAD
TCM1135-025G TCM1135-500G
3-Methoxydiphenylamine ≥98.0%
3-Methoxydiphenylamine

CAS Number: 101-16-6
MDL Number: MFCD00008383
Molecular Formula: C13H13NO
Molecular Weight: 199.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 160
Melting point (°C): 71

Formula: C₁₃H₁₃NO
MW: 199.25 g/mol
Boiling Pt: 160 °C
Melting Pt: 71 °C
MDL Number: MFCD00008383
CAS Number: 101-16-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a97085201aa74bde4670570462cb166c76d35866&vid=a97085201aa74bde4670570462cb166c76d35866&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR