2,2'-Methylenebis(4-methylphenol) ≥90.0%

Supplier: TCI America

Synonyms: 2,2'-Methylenebis(p-cresol), 2,2'-Dihydroxy-5,5'-dimethyldiphenylmethane, Bis(2-hydroxy-5-methylphenyl)methane

M1134-25G M1134-5G
TCM1134-025GEA 340.65 CAD
TCM1134-025G 76220-230
2,2'-Methylenebis(4-methylphenol) ≥90.0%
2,2'-Methylenebis(4-methylphenol)

CAS Number: 3236-63-3
MDL Number: MFCD00155176
Molecular Formula: C15H16O2
Molecular Weight: 228.29
Purity/Analysis Method: >90.0% (GC)
Form: Crystal
Melting point (°C): 125
Flash Point (°C): 218


Product Categories: Materials Chemistry, Monomers, Diphenylmethanes (for High-Performance Polymer Research)
Other Product Names / Synonyms: Bis(2-hydroxy-5-methylphenyl)methane, 2,2'-Dihydroxy-5,5'-dimethyldiphenylmethane, 2,2'-Methylenebis(p-cresol)

Formula: C₁₅H₁₆O₂
MW: 228.29 g/mol
Melting Pt: 125 °C
Flash Pt: 218 °C
MDL Number: MFCD00155176
CAS Number: 3236-63-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7f4c737212d22c87a5c48025d4f3761096ed1b6&vid=c7f4c737212d22c87a5c48025d4f3761096ed1b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR