Ethyl-2-methoxy-6-methylbenzoate ≥95.0%

Supplier: TCI America

Synonyms: Ethyl 2-methoxy-6-methylbenzoate, 2-Methoxy-6-methylbenzoic Acid Ethyl Ester

M1120-5G M1120-25G
TCM1120-005GEA 385.2 CAD
TCM1120-005G TCM1120-25G
Ethyl-2-methoxy-6-methylbenzoate ≥95.0%
Ethyl-2-methoxy-6-methylbenzoate

CAS Number: 6520-83-8
MDL Number: MFCD00191666
Molecular Formula: C11H14O3
Molecular Weight: 194.23
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: 2-Methoxy-6-methylbenzoic Acid Ethyl Ester

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 135 °C
Density: 1.09 (20 °C)
MDL Number: MFCD00191666
CAS Number: 6520-83-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a98b03944d6c07004ff4e4e135138908760802c3&vid=a98b03944d6c07004ff4e4e135138908760802c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR