4-Acetamido-1-benzylpiperidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1654-5G A1654-25G
TCA1654-5GEA 123.24 CAD
TCA1654-5G TCA1654-25G
4-Acetamido-1-benzylpiperidine ≥98.0% (by GC, titration analysis)
4-Acetamido-1-benzylpiperidine

CAS Number: 50534-23-1
MDL Number: MFCD00247343
Molecular Formula: C14H20N2O
Molecular Weight: 232.33
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 140

Formula: C₁₄H₂₀N₂O
Melting Pt: 140 °C
MDL Number: MFCD00247343
CAS Number: 50534-23-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f04e4dd7bdb9528bc26f3851c2fc0eafd32faf2&vid=7f04e4dd7bdb9528bc26f3851c2fc0eafd32faf2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR