Melatonin ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

M1105-5G M1105-1G
TCM1105-005GEA 227.52 CAD
TCM1105-005G TCM1105-001G
Melatonin ≥98.0% (by HPLC, total nitrogen)
Melatonine

CAS Number: 73-31-4
MDL Number: MFCD00005655
Molecular Formula: C13H16N2O2
Molecular Weight: 232.28
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 118


Product Categories: Bioscience, Antioxidants, Other Pharmaceutical Ingredients, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: N-Acetyl-5-methoxytryptamine

Formula: C₁₃H₁₆N₂O₂
Melting Pt: 118 °C
MDL Number: MFCD00005655
CAS Number: 73-31-4
EINECS: 200-797-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29b2ba04965e75f1033704841229497d0e17c5a3&vid=29b2ba04965e75f1033704841229497d0e17c5a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR