4,4'-Methylenebis(2,6-dimethylphenol) ≥98.0%

Supplier: TCI America

Synonyms: Bis(4-hydroxy-3,5-dimethylphenyl)methane, 4,4'-Dihydroxy-3,3',5,5'-tetramethyldiphenylmethane, 2,2',6,6'-Tetramethyl-4,4'-methylenediphenol

M1099-25G M1099-500G
TCM1099-025GEA 202.24 CAD
TCM1099-025G TCM1099-500G
4,4'-Methylenebis(2,6-dimethylphenol) ≥98.0%
4,4'-Methylenebis(2,6-dimethylphenol)

CAS Number: 5384-21-4
MDL Number: MFCD00055640
Molecular Formula: C17H20O2
Molecular Weight: 256.35
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 176


Product Categories: Materials Chemistry, Monomers, Diphenylmethanes (for High-Performance Polymer Research)
Other Product Names / Synonyms: Bis(4-hydroxy-3,5-dimethylphenyl)methane, 4,4'-Dihydroxy-3,3',5,5'-tetramethyldiphenylmethane, 2,2',6,6'-Tetramethyl-4,4'-methylenediphenol

Formula: C₁₇H₂₀O₂
MW: 256.35 g/mol
Melting Pt: 176 °C
MDL Number: MFCD00055640
CAS Number: 5384-21-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39d72481de27545ebf843143502231841e89262e&vid=39d72481de27545ebf843143502231841e89262e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR