(S)-2-Aminopent-4-enoic acid hydrochloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

A1648-1G A1648-200MG A1648-5G
TCA1648-1GEA 285.98 CAD
TCA1648-1G TCA1648-200MG TCA1648-5G
(S)-2-Aminopent-4-enoic acid hydrochloride ≥97.0% (by titrimetric analysis)
(S)-2-Aminopent-4-enoic acid hydrochloride

CAS Number: 195316-72-4
MDL Number: MFCD06797042
Molecular Formula: C5H9NO2
Molecular Weight: 151.59
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Specific rotation [a]20/D: -10 deg (C=2, H2O)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-2-Amino-4-pentenoic Acid Hydrochloride

Formula: C₅H₁₀ClNO₂
MDL Number: MFCD00002627
CAS Number: 195316-72-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d83c0ca72214990dfd8b7ccff3bfc5b8b9d2297&vid=8d83c0ca72214990dfd8b7ccff3bfc5b8b9d2297&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR