4,4'-(ɑ-Methylbenzylidene)bisphenol ≥98.0%

Supplier: TCI America

M1098-25G M1098-500G
TCM1098-025GEA 112.18 CAD
TCM1098-025G TCM1098-500G
4,4'-(ɑ-Methylbenzylidene)bisphenol ≥98.0%
4,4'-(alpha-Methylbenzylidene)bisphenol

CAS Number: 1571-75-1
MDL Number: MFCD00134687
Molecular Formula: C20H18O2
Molecular Weight: 290.36
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 189
Flash Point (°C): 250


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,1-Bis(4-hydroxyphenyl)-1-phenylethane, 4,4'-(1-Phenylethylidene)diphenol

Formula: C₂₀H₁₈O₂
Melting Pt: 189 °C
Flash Pt: 250 °C
MDL Number: MFCD00134687
CAS Number: 1571-75-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a92d86c942f7bbe88dd7ed0268f152a3a5fa244&vid=2a92d86c942f7bbe88dd7ed0268f152a3a5fa244&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR