tert-Butyl methyl(pyrrolidin-3-yl)carbamate ≥96.0%

Supplier: TCI America

M1096-5G M1096-25G
TCM1096-005GEA 271.22 CAD
TCM1096-005G TCM1096-25G
tert-Butyl methyl(pyrrolidin-3-yl)carbamate ≥96.0%
tert-Butyl methyl(pyrrolidin-3-yl)carbamate

CAS Number: 172478-00-1
MDL Number: MFCD00191665
Molecular Formula: C10H20N2O2
Molecular Weight: 200.28
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: 3-(N-Boc-N-methylamino)pyrrolidine

Formula: C₁₀H₂₀N₂O₂
Boiling Pt: 96 °C
Density: 1.02 (20 °C)
MDL Number: MFCD00191665
CAS Number: 172478-00-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b4e8291d2a5424ce2785aac7cfb91de27344536&vid=3b4e8291d2a5424ce2785aac7cfb91de27344536&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR