2-Mercapto-4(3H)-quinazolinone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-Mercaptoquinazolin-4(3H)-one, 2-Thio-4(3H)-quinazolinone

M1089-5G M1089-25G
TCM1089-005GEA 211.72 CAD
TCM1089-005G TCM1089-25G
2-Mercapto-4(3H)-quinazolinone ≥98.0% (by HPLC, titration analysis)
2-Mercapto-4(3H)-quinazolinone

CAS Number: 13906-09-7
MDL Number: MFCD00006885
Molecular Formula: C8H6N2OS
Molecular Weight: 178.21
Purity/Analysis Method: >98.0% (T,HPLC)
Form: Crystal


Other Product Names / Synonyms: 2-Thio-4(3H)-quinazolinone

Formula: C₈H₆N₂OS
MW: 178.21 g/mol
MDL Number: MFCD00006885
CAS Number: 13906-09-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5afb67552e4e0edd7fb4001e38b540969822dacc&vid=5afb67552e4e0edd7fb4001e38b540969822dacc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T,HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR