8-Methoxyquinaldine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M1088-5G M1088-1G
TCM1088-5GEA 439.71 CAD
TCM1088-5G TCM1088-001G
8-Methoxyquinaldine ≥98.0% (by GC, titration analysis)
8-Methoxyquinaldine

CAS Number: 3033-80-5
MDL Number: MFCD00034436
Molecular Formula: C11H11NO
Molecular Weight: 173.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 124


Other Product Names / Synonyms: 8-Methoxyquinaldine

Formula: C₁₁H₁₁NO
Melting Pt: 124 °C
MDL Number: MFCD00034436
CAS Number: 3033-80-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f6595b311e0d62d9a79293f3bcd10806277dde5&vid=5f6595b311e0d62d9a79293f3bcd10806277dde5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR