6-Methoxy-4-methylquinoline hydrate ≥98.0%

Supplier: TCI America

Synonyms: 6-Methoxy-4-methylquinoline

M1087-5G M1087-1G
TCM1087-5GEA 841.35 CAD
TCM1087-5G TCM1087-001G
6-Methoxy-4-methylquinoline hydrate ≥98.0%
6-Methoxy-4-methylquinoline hydrate

CAS Number: 41037-26-7
MDL Number: MFCD00191547
Molecular Formula: C11H11NO
Molecular Weight: 173.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 158
Melting point (°C): 53

Formula: C₁₁H₁₁NO
MW: 173.22 g/mol
Boiling Pt: 158 °C
Melting Pt: 53 °C
MDL Number: MFCD00191547
CAS Number: 41037-26-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08506f0dbb02bf5afa739ba22a1e3e4254813ed0&vid=08506f0dbb02bf5afa739ba22a1e3e4254813ed0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR