2-Ethylhexyl-4-methoxycinnamate ≥97.0%

Supplier: TCI America

Synonyms: 4-Methoxycinnamic Acid 2-Ethylhexyl Ester, 4-Methoxycinnamic Acid Octyl Ester, Octinoxate, Octyl 4-Methoxycinnamate, 2-Ethylhexyl 3-(4-methoxyphenyl)acrylate

M1082-25ML M1082-500ML M1082-100ML
TCM1082-025MLEA 72.32 CAD
TCM1082-025ML TCM1082-500ML TCM1082-100ML
2-Ethylhexyl-4-methoxycinnamate ≥97.0%
2-Ethylhexyl-4-methoxycinnamate

CAS Number: 5466-77-3
MDL Number: MFCD00072582
Molecular Formula: C18H26O3
Molecular Weight: 290.40
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 193
Specific Gravity (20/20): 1.01


Product Categories: Bioscience, Cosmetic Ingredients for Research and Experimental Use, Phenylpropanoids
Other Product Names / Synonyms: 4-Methoxycinnamic Acid 2-Ethylhexyl Ester, 4-Methoxycinnamic Acid Octyl Ester, Octyl 4-Methoxycinnamate

Formula: C₁₈H₂₆O₃
MW: 290.40 g/mol
Density: 1.01 (20 °C)
Flash Pt: 193 °C
MDL Number: MFCD00072582
CAS Number: 5466-77-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=465ef7f7e693296de54d81654b6069a1e66cc88c&vid=465ef7f7e693296de54d81654b6069a1e66cc88c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR