Monocaprin ≥98.0%
Supplier: TCI America
M1072-1G
M1072-5G
TCM1072-001GEA
192.76
CAD
TCM1072-001G
TCM1072-005G
Monocaprin ≥98.0%
Monocaprin
CAS Number: 26402-22-2
MDL Number: MFCD00056656
Molecular Formula: C13H26O4
Molecular Weight: 246.35
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 53
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Glycerol alpha-Monodecanoate, 1-Monodecanoyl Glycerol
Formula:
C₁₃H₂₆O₄ Melting Pt: 53 °C |
MDL Number:
MFCD00056656 CAS Number: 26402-22-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...