2-Ethylhexyl-3-mercaptopropionate ≥98.0%

Supplier: TCI America

Synonyms: 3-Mercaptopropionic Acid 2-Ethylhexyl Ester, 2-Ethylhexyl 3-mercaptopropanoate, 2-Octyl 3-Mercaptopropionate, 3-Mercaptopropionic Acid Octyl Ester

M1067-25ML M1067-500ML
TCM1067-025MLEA 62.88 CAD
TCM1067-025ML TCM1067-500ML
2-Ethylhexyl-3-mercaptopropionate ≥98.0%
2-Ethylhexyl-3-mercaptopropionate

CAS Number: 50448-95-8
MDL Number: MFCD00087933
Molecular Formula: C11H22O2S
Molecular Weight: 218.36
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 125
Specific Gravity (20/20): 0.96


Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents
Other Product Names / Synonyms: 3-Mercaptopropionic Acid 2-Ethylhexyl Ester, 3-Mercaptopropionic Acid Octyl Ester, 2-Octyl 3-Mercaptopropionate

Formula: C₁₁H₂₂O₂S
MW: 218.36 g/mol
Density: 0.96 (20 °C)
Flash Pt: 125 °C
MDL Number: MFCD00087933
CAS Number: 50448-95-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a784db9c3bb7d9ddb9dc8f5f68b340e23761f9c&vid=4a784db9c3bb7d9ddb9dc8f5f68b340e23761f9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR