(R)-(+)-Menthyltoluene-4-sulfinate ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: (1S,2R,5S)-(+)-Menthyl (R)-p-Toluenesulfinate, (1S,2R,5S)-(+)-Menthyl (R)-p-Toluenesulphinate

M1066-1G M1066-5G
TCM1066-001GEA 66.36 CAD
TCM1066-001G TCM1066-5G
(R)-(+)-Menthyltoluene-4-sulfinate ≥97.0% (by HPLC)
(R)-(+)-Menthyltoluene-4-sulphinate

CAS Number: 91796-57-5
MDL Number: MFCD00064472
Molecular Formula: C17H26O2S
Molecular Weight: 294.45
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 103
Specific rotation [a]20/D: 200 deg (C=2, Acetone)
Storage Temperature: <0°C

Formula: C₁₇H₂₆O₂S
Melting Pt: 103 °C
MDL Number: MFCD00064472
CAS Number: 91796-57-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba4772ccd0a53123e8e18dcb7f9f11b2786e2b53&vid=ba4772ccd0a53123e8e18dcb7f9f11b2786e2b53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR