1-(4-Methylphenoxy)-2-propanol ≥90.0%

Supplier: TCI America

M1065-25G M1065-5G
TCM1065-025GEA 416.49 CAD
TCM1065-025G TCM1065-005G
1-(4-Methylphenoxy)-2-propanol ≥90.0%
1-(4-Methylphenoxy)-2-propanol

CAS Number: 4369-08-8
MDL Number: MFCD00191544
Molecular Formula: C10H14O2
Molecular Weight: 166.22
Purity/Analysis Method: >90.0% (GC)
Form: Crystal
Boiling point (°C): 130
Freezing point (°C): 28


Other Product Names / Synonyms: 1-(p-Tolyloxy)-2-propanol

Formula: C₁₀H₁₄O₂
Boiling Pt: 130 °C
MDL Number: MFCD00191544
CAS Number: 4369-08-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fecc79974a795ce3953b7d15d1bbb81fafceaf3&vid=9fecc79974a795ce3953b7d15d1bbb81fafceaf3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR