β-Methyllevulinic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M1057-5G
M1057-25G
TCM1057-005GEA
515.71
CAD
TCM1057-005G
TCM1057-25G
β-Methyllevulinic acid ≥98.0% (by GC, titration analysis)
beta-Methyllevulinic acid
CAS Number: 6628-79-1
MDL Number: MFCD00143691
Molecular Formula: C6H10O3
Molecular Weight: 130.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 242
Melting point (°C): 32
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 3-Acetylbutyric Acid, 3-Methyl-4-oxopentanoic Acid
Formula:
C₆H₁₀O₃ Boiling Pt: 242 °C Melting Pt: 32 °C |
MDL Number:
MFCD00143691 CAS Number: 6628-79-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...