trans-1-Methyl-4-carboxy-5-(3-pyridyl)-2-pyrrolidinone ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: trans-4'-Carboxycotinine

M1053-5G M1053-100MG M1053-1G
TCM1053-5GEA 255.96 CAD
TCM1053-5G TCM1053-100MG TCM1053-1G
trans-1-Methyl-4-carboxy-5-(3-pyridyl)-2-pyrrolidinone ≥95.0% (by GC, titration analysis)
trans-1-Methyl-4-carboxy-5-(3-pyridyl)-2-pyrrolidinone

CAS Number: 33224-01-0
MDL Number: MFCD00075394
Molecular Formula: C11H12N2O3
Molecular Weight: 220.23
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 194


Other Product Names / Synonyms: trans-4'-Carboxycotinine

Formula: C₁₁H₁₂N₂O₃
MW: 220.23 g/mol
Melting Pt: 194 °C
MDL Number: MFCD00075394
CAS Number: 33224-01-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fc1876290be3cfed3ace6fc6ce906eab0a45ed3&vid=8fc1876290be3cfed3ace6fc6ce906eab0a45ed3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR