N-Methyl-3-chloropropylamine hydrochloride ≥99.0%

Supplier: TCI America

Synonyms: 3-(Methylamino)propyl Chloride Hydrochloride, 3-Chloro-N-methylpropan-1-amine hydrochloride

M1048-1G M1048-5G
TCM1048-001GEA 203.35 CAD
TCM1048-001G TCM1048-005G
N-Methyl-3-chloropropylamine hydrochloride ≥99.0%
N-Methyl-3-chloropropylamine hydrochloride

CAS Number: 97145-88-5
MDL Number: MFCD00144935
Molecular Formula: C4H10ClN
Molecular Weight: 144.04
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 105


Other Product Names / Synonyms: 3-(Methylamino)propyl Chloride Hydrochloride

Formula: C₄H₁₀ClN
MW: 144.04 g/mol
Melting Pt: 105 °C
MDL Number: MFCD00144935
CAS Number: 97145-88-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64c573539af6546b0d033aae839be5a089ca6d7a&vid=64c573539af6546b0d033aae839be5a089ca6d7a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR